Assay Details
| Gene Symbol: Yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009712.1 | NP_001009712.1 | 1 - 2 | 282 | 137 | |
| GenBank mRNA | BC088468.1 | - | 1 - 2 | 282 | 7127993 | 137 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC088468.1, NM_001009712.1 transcripts.
Set Membership: |
|

Yipf4
Genomic Map