Assay Details
| Gene Symbol: Galm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007704.1 | NP_001007705.1 | 6 - 7 | 1007 | 115 | |
| GenBank mRNA | BC081876.1 | - | 6 - 7 | 1007 | 7107751 | 115 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P840715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95814 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P840715 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P840715 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P840715 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P546279 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P840715 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27705 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2280 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46490 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14423 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 105022 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10882500 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882497 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882498 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882499 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882503 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882502 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882501 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10882496 | - | - |
| Rat Genome 230 2.0 Array | 1385072_at | - | - |
| Rat Genome 230 2.0 Array | 1385072_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379250_at | - | - |
| Rat Genome 230 2.0 Array | 1379250_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379250_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385072_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385072_at | - | - |
| Affymetrix HT Rat Focus Array | 1379250_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600000260.rn.1 | TC0600000038.rn.1 | RTA TC |
| Rat Expression Set 230 | 1379250_at | - | - |
| Rat Expression Set 230 | 1379250_PM_at | - | - |
| Rat Expression Set 230 | 1385072_PM_at | - | - |
| Rat Expression Set 230 | 1385072_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10882496 | - | - |
| Rat Genome U34 Set | rc_AA892666_at | - | - |
| Rat Genome U34 Set | rc_AA997048_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21696589 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363661 | - | - |
| RatRef-12 Expression BeadChip | ILMN_64854 | - | - |

Galm
Genomic Map