Assay Details
| Gene Symbol: Sdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013026.2 | NP_037158.1 | 2 - 3 | 359 | 131 | |
| GenBank mRNA | BC061834.1 | - | 2 - 3 | 359 | 5600086 | 131 |
| M81785.1 | - | 2 - 3 | 374 | 131 | ||
| S61865.1 | - | 2 - 3 | 374 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sdc1
Genomic Map