Assay Details
| Gene Symbol: LOC690035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764722.1 | XP_008762944.1 | 16 - 17 | 3097 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690035| Gene Symbol: LOC690035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008764722.1 | XP_008762944.1 | 16 - 17 | 3097 | 115 | |
Set Membership: |
|