Assay Details
Gene Symbol: LOC690035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764722.1 | XP_008762944.1 | 21 - 22 | 3805 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
