Assay Details
Gene Symbol: Mgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246184.2 | XP_006246246.1 | 2 - 3 | 792 | 104 | |
XM_006246185.2 | XP_006246247.1 | 3 - 4 | 618 | 104 | ||
GenBank mRNA | AB012877.1 | - | 2 - 3 | 157 | 104 | |
AB012878.1 | - | 2 - 3 | 195 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006246184.2, XM_006246185.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
