Assay Details
| Gene Symbol: Ube2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031138.2 | NP_112400.1 | 1 - 2 | 146 | 93 | |
| XM_006246343.2 | XP_006246405.1 | 2 - 3 | 171 | 93 | ||
| GenBank mRNA | BC070946.1 | - | 1 - 2 | 139 | 7103580 | 93 |
| DN931313.1 | - | 1 - 2 | 176 | 93 | ||
| FQ222428.1 | - | 1 - 2 | 163 | 93 | ||
| M62388.1 | - | 1 - 2 | 88 | 93 | ||
| Gene Symbol: LOC103694902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773893.1 | XP_008772115.1 | 2 - 3 | 170 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Ube2b
Genomic Map