Assay Details
Gene Symbol: Med1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134361.1 | NP_001127833.1 | 10 - 11 | 810 | 63 | |
XM_006247398.2 | XP_006247460.1 | 10 - 11 | 960 | 63 | ||
XM_006247399.2 | XP_006247461.1 | 11 - 12 | 1035 | 63 | ||
XM_006247401.2 | XP_006247463.1 | 10 - 11 | 959 | 63 | ||
XM_008768105.1 | XP_008766327.1 | 10 - 11 | 1590 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
cell morphogenesis
angiogenesis
in utero embryonic development
liver development
embryonic placenta development
lens development in camera-type eye
thyroid hormone mediated signaling pathway
ventricular trabecula myocardium morphogenesis
retinal pigment epithelium development
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase I promoter
transcription initiation from RNA polymerase II promoter
thyroid hormone generation
androgen biosynthetic process
multicellular organism development
brain development
heart development
lactation
positive regulation of cell proliferation
organ morphogenesis
positive regulation of gene expression
negative regulation of keratinocyte proliferation
protein ubiquitination
keratinocyte differentiation
monocyte differentiation
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
organ regeneration
positive regulation of intracellular estrogen receptor signaling pathway
positive regulation of protein import into nucleus, translocation
mammary gland epithelial cell proliferation
positive regulation of mammary gland epithelial cell proliferation
embryonic heart tube development
embryonic hindlimb morphogenesis
embryonic hemopoiesis
peroxisome proliferator activated receptor signaling pathway
cellular response to hepatocyte growth factor stimulus
megakaryocyte development
mRNA transcription from RNA polymerase II promoter
camera-type eye development
negative regulation of apoptotic process
fat cell differentiation
positive regulation of keratinocyte differentiation
negative regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
erythrocyte development
enucleate erythrocyte development
positive regulation of interferon-gamma-mediated signaling pathway
mammary gland branching involved in thelarche
mammary gland branching involved in pregnancy
epithelial cell proliferation involved in mammary gland duct elongation
positive regulation of G0 to G1 transition
ERK1 and ERK2 cascade
regulation of vitamin D receptor signaling pathway
cellular response to epidermal growth factor stimulus
cellular response to steroid hormone stimulus
cellular response to thyroid hormone stimulus
positive regulation of receptor activity
positive regulation of hepatocyte proliferation
regulation of RNA biosynthetic process
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter binding
RNA polymerase II transcription cofactor activity
DNA binding
chromatin binding
transcription cofactor activity
transcription coactivator activity
receptor activity
transcription factor binding
ligand-dependent nuclear receptor binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
thyroid hormone receptor coactivator activity
chromatin DNA binding
protein complex binding
nuclear hormone receptor binding
mediator complex binding
vitamin D receptor binding
retinoic acid receptor binding
peroxisome proliferator activated receptor binding
thyroid hormone receptor binding
LBD domain binding
ubiquitin protein ligase activity
negative regulation of transcription from RNA polymerase II promoter
cell morphogenesis
angiogenesis
in utero embryonic development
liver development
embryonic placenta development
lens development in camera-type eye
thyroid hormone mediated signaling pathway
ventricular trabecula myocardium morphogenesis
retinal pigment epithelium development
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase I promoter
transcription initiation from RNA polymerase II promoter
thyroid hormone generation
androgen biosynthetic process
multicellular organism development
brain development
heart development
lactation
positive regulation of cell proliferation
organ morphogenesis
positive regulation of gene expression
negative regulation of keratinocyte proliferation
protein ubiquitination
keratinocyte differentiation
monocyte differentiation
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
organ regeneration
positive regulation of intracellular estrogen receptor signaling pathway
positive regulation of protein import into nucleus, translocation
mammary gland epithelial cell proliferation
positive regulation of mammary gland epithelial cell proliferation
embryonic heart tube development
embryonic hindlimb morphogenesis
embryonic hemopoiesis
peroxisome proliferator activated receptor signaling pathway
cellular response to hepatocyte growth factor stimulus
megakaryocyte development
mRNA transcription from RNA polymerase II promoter
camera-type eye development
negative regulation of apoptotic process
fat cell differentiation
positive regulation of keratinocyte differentiation
negative regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
erythrocyte development
enucleate erythrocyte development
positive regulation of interferon-gamma-mediated signaling pathway
mammary gland branching involved in thelarche
mammary gland branching involved in pregnancy
epithelial cell proliferation involved in mammary gland duct elongation
positive regulation of G0 to G1 transition
ERK1 and ERK2 cascade
regulation of vitamin D receptor signaling pathway
cellular response to epidermal growth factor stimulus
cellular response to steroid hormone stimulus
cellular response to thyroid hormone stimulus
positive regulation of receptor activity
positive regulation of hepatocyte proliferation
regulation of RNA biosynthetic process
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter binding
RNA polymerase II transcription cofactor activity
DNA binding
chromatin binding
transcription cofactor activity
transcription coactivator activity
receptor activity
transcription factor binding
ligand-dependent nuclear receptor binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
thyroid hormone receptor coactivator activity
chromatin DNA binding
protein complex binding
nuclear hormone receptor binding
mediator complex binding
vitamin D receptor binding
retinoic acid receptor binding
peroxisome proliferator activated receptor binding
thyroid hormone receptor binding
LBD domain binding
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P838798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170663 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P838798 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P838798 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P838798 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17680 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18455 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P838798 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10746889 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746876 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746884 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746890 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746875 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746887 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746882 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746880 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746881 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746878 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746886 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746891 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746883 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746885 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746888 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746879 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746877 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10746872 | - | - |
Rat Genome 230 2.0 Array | 1376906_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390506_at | - | - |
Rat Genome 230 2.0 Array | 1384962_at | - | - |
Rat Genome 230 2.0 Array | 1376906_at | - | - |
Rat Genome 230 2.0 Array | 1384962_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376582_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390506_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376582_at | - | - |
Affymetrix HT Rat Focus Array | 1390506_at | - | - |
Affymetrix HT Rat Focus Array | 1376906_at | - | - |
Affymetrix HT Rat Focus Array | 1376906_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390506_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376582_at | - | - |
Affymetrix HT Rat Focus Array | 1384962_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1384962_at | - | - |
Affymetrix HT Rat Focus Array | 1376582_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000033156.rn.1 | TC1000003705.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000033157.rn.1 | TC1000003705.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000033158.rn.1 | TC1000003705.rn.1 | RTA TC |
Rat Expression Set 230 | 1390506_PM_at | - | - |
Rat Expression Set 230 | 1376582_at | - | - |
Rat Expression Set 230 | 1390506_at | - | - |
Rat Expression Set 230 | 1376906_PM_at | - | - |
Rat Expression Set 230 | 1376906_at | - | - |
Rat Expression Set 230 | 1376582_PM_at | - | - |
Rat Expression Set 230 | 1384962_at | - | - |
Rat Expression Set 230 | 1384962_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10746872 | - | - |
Rat Genome U34 Set | rc_AA800817_at | - | - |
Rat Genome U34 Set | rc_AI060320_at | - | - |
Rat Genome U34 Set | rc_AA997858_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21133595 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354875 | - | - |
