Assay Details
| Gene Symbol: Tbc1d24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105769.1 | NP_001099239.1 | 4 - 5 | 1400 | 61 | |
| XM_006245926.1 | XP_006245988.1 | 3 - 4 | 1159 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d24
Genomic Map