Assay Details
| Gene Symbol: Trim16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135033.2 | NP_001128505.1 | 2 - 3 | 634 | 63 | |
| XM_008767824.1 | XP_008766046.1 | 2 - 3 | 895 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trim16
Genomic Map