Assay Details
Gene Symbol: Rangrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105790.1 | NP_001099260.1 | 1 - 2 | 212 | 103 | |
GenBank mRNA | FQ228156.1 | - | 1 - 2 | 214 | 103 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
