Assay Details
| Gene Symbol: Aoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031582.2 | NP_113770.2 | 1 | 939 | 83 | |
| GenBank mRNA | AB195675.1 | - | 1 | 944 | 83 | |
| BC100613.1 | - | 1 | 939 | 5598211 | 83 | |
| U72632.1 | - | 1 | 944 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Aoc3
Genomic Map