Assay Details
| Gene Symbol: Mmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007673.1 | NP_001007674.1 | 5 - 6 | 638 | 89 | |
| XM_006247126.1 | XP_006247188.1 | 5 - 6 | 676 | 89 | ||
| XM_006247127.2 | XP_006247189.1 | 5 - 6 | 1498 | 89 | ||
| GenBank mRNA | AF540876.1 | - | 5 - 6 | 537 | 89 | |
| BC081965.1 | - | 5 - 6 | 638 | 7125235 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmd
Genomic Map