Assay Details
Gene Symbol: Smim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277479.1 | NP_001264408.1 | 1 - 2 | 164 | 110 | |
GenBank mRNA | CB736773.1 | - | 1 - 2 | 164 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277479.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
