Assay Details
| Gene Symbol: Gps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017477.1 | NP_001017477.1 | 4 - 5 | 680 | 111 | |
| XM_006246783.2 | XP_006246845.1 | 5 - 6 | 630 | 111 | ||
| XM_006246785.1 | XP_006246847.1 | 5 - 6 | 630 | 111 | ||
| XM_008767855.1 | XP_008766077.1 | 5 - 6 | 630 | 111 | ||
| XM_008767856.1 | XP_008766078.1 | 5 - 6 | 630 | 111 | ||
| GenBank mRNA | BC083699.1 | - | 4 - 5 | 680 | 7190067 | 111 |
| FQ233878.1 | - | 5 - 6 | 513 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gps2
Genomic Map