Assay Details
| Gene Symbol: Gps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017477.1 | NP_001017477.1 | 8 - 9 | 1090 | 72 | |
| XM_006246783.2 | XP_006246845.1 | 9 - 10 | 1040 | 72 | ||
| XM_006246785.1 | XP_006246847.1 | 9 - 10 | 1040 | 72 | ||
| XM_008767855.1 | XP_008766077.1 | 8 - 9 | 1115 | 72 | ||
| XM_008767856.1 | XP_008766078.1 | 8 - 9 | 1115 | 72 | ||
| GenBank mRNA | BC083699.1 | - | 8 - 9 | 1090 | 7190067 | 72 |
| FQ233878.1 | - | 9 - 10 | 924 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gps2
Genomic Map