Assay Details
| Gene Symbol: Cd300e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166577.1 | NP_001160049.1 | 2 - 3 | 460 | 75 | |
| XM_008768394.1 | XP_008766616.1 | 3 - 4 | 786 | 75 | ||
| XM_008768395.1 | XP_008766617.1 | 3 - 4 | 790 | 75 | ||
| GenBank mRNA | GU057983.1 | - | 2 - 3 | 460 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd300e
Genomic Map