Assay Details
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106711.1 | NP_001100181.1 | 1 - 2 | 175 | 57 | |
XM_006239798.2 | XP_006239860.1 | 1 - 2 | 298 | 57 | ||
GenBank mRNA | BC166997.1 | - | 1 - 2 | 216 | 8365241 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
