Assay Details
| Gene Symbol: Mboat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108016.2 | NP_001101486.2 | 2 - 3 | 332 | 79 | |
| XM_006239985.2 | XP_006240047.1 | 2 - 3 | 302 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mboat2
Genomic Map