Assay Details
| Gene Symbol: Trappc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106733.1 | NP_001100203.1 | 3 - 4 | 439 | 146 | |
| GenBank mRNA | FQ230024.1 | - | 3 - 4 | 202 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trappc6b
Genomic Map