Assay Details
| Gene Symbol: Fam49a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106718.2 | NP_001100188.1 | 11 - 12 | 1101 | 76 | |
| XM_008764584.1 | XP_008762806.1 | 11 - 12 | 1650 | 76 | ||
| XM_008764585.1 | XP_008762807.1 | 10 - 11 | 1003 | 76 | ||
| GenBank mRNA | BC158700.1 | - | 11 - 12 | 1101 | 9028479 | 76 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P547629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146401 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P547629 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P547629 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P547629 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1007965 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1569 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18035 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6330 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10883645 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883637 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883640 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883648 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883647 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883646 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883642 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883639 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883643 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883644 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883638 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883641 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10883636 | - | - |
| Rat Genome 230 2.0 Array | 1394470_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1394470_at | - | - |
| Affymetrix HT Rat Focus Array | 1394470_at | - | - |
| Affymetrix HT Rat Focus Array | 1394470_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600002960.rn.1 | TC0600000435.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002958.rn.1 | TC0600000435.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002959.rn.1 | TC0600000435.rn.1 | RTA TC |
| Rat Expression Set 230 | 1394470_at | - | - |
| Rat Expression Set 230 | 1394470_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883636 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21087774 | - | - |
| ABI Rat Genome Survey Microarray | 21703445 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355101 | - | - |

Fam49a
Genomic Map