Assay Details
| Gene Symbol: Iah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100540.1 | NP_001094010.1 | 4 - 5 | 457 | 60 | |
| GenBank mRNA | BC168771.1 | - | 4 - 5 | 457 | 6919650 | 60 |
| FQ220941.1 | - | 4 - 5 | 453 | 60 | ||
| FQ225724.1 | - | 4 - 5 | 482 | 60 | ||
| FQ231316.1 | - | 4 - 5 | 482 | 60 | ||
| FQ232084.1 | - | 4 - 5 | 487 | 60 | ||
| FQ235324.1 | - | 4 - 5 | 482 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Iah1
Genomic Map