Assay Details
| Gene Symbol: Pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004442.1 | NP_001004442.1 | 7 - 8 | 730 | 83 | |
| GenBank mRNA | BC082063.1 | - | 7 - 8 | 730 | 7132589 | 83 |
| X79328.1 | - | 6 - 7 | 670 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdia6
Genomic Map