Assay Details
| Gene Symbol: Nfkbia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105720.2 | NP_001099190.2 | 3 - 4 | 667 | 108 | |
| GenBank mRNA | BC166886.1 | - | 3 - 4 | 619 | 6890357 | 108 |
| CB316505.1 | - | 3 - 4 | 621 | 108 | ||
| FQ214065.1 | - | 1 - 2 | 115 | 108 | ||
| FQ215246.1 | - | 3 - 4 | 619 | 108 | ||
| FQ225350.1 | - | 3 - 4 | 614 | 108 | ||
| FQ225877.1 | - | 3 - 4 | 617 | 108 | ||
| FQ227283.1 | - | 3 - 4 | 617 | 108 | ||
| FQ233200.1 | - | 3 - 4 | 617 | 108 | ||
| FQ233557.1 | - | 3 - 4 | 619 | 108 | ||
| FQ233627.1 | - | 3 - 4 | 605 | 108 | ||
| FQ234262.1 | - | 3 - 4 | 619 | 108 | ||
| X63594.1 | - | 3 - 4 | 597 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfkbia
Genomic Map