Assay Details
Gene Symbol: Rlim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257149.2 | XP_006257211.1 | 2 - 3 | 270 | 104 |
Gene Symbol: LOC100910646 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257141.2 | XP_006257203.1 | 270 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006257141.2, XM_006257149.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
