Assay Details
| Gene Symbol: Magee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079891.1 | NP_001073360.1 | 1 | 3241 | 150 | |
| GenBank mRNA | BC128754.1 | - | 1 | 3241 | 7103930 | 150 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Magee1
Genomic Map