Assay Details
| Gene Symbol: Aldh3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006998.1 | NP_001006999.1 | 8 - 9 | 1152 | 57 | |
| XM_006230787.2 | XP_006230849.1 | 9 - 10 | 1455 | 57 | ||
| XM_008760132.1 | XP_008758354.1 | 8 - 9 | 1642 | 57 | ||
| XM_008760133.1 | XP_008758355.1 | 8 - 9 | 1642 | 57 | ||
| XM_008760134.1 | XP_008758356.1 | 8 - 9 | 1136 | 57 | ||
| GenBank mRNA | BC083850.1 | - | 8 - 9 | 1152 | 7122451 | 57 |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh3b1
Genomic Map