Assay Details
Gene Symbol: Aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006998.1 | NP_001006999.1 | 4 - 5 | 429 | 92 | |
XM_006230787.2 | XP_006230849.1 | 5 - 6 | 732 | 92 | ||
XM_008760132.1 | XP_008758354.1 | 4 - 5 | 919 | 92 | ||
XM_008760133.1 | XP_008758355.1 | 4 - 5 | 919 | 92 | ||
XM_008760134.1 | XP_008758356.1 | 4 - 5 | 413 | 92 | ||
GenBank mRNA | BC083850.1 | - | 4 - 5 | 429 | 7122451 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
