Assay Details
| Gene Symbol: Mrpl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107567.1 | NP_001101037.1 | 1 - 2 | 266 | 81 | |
| XM_008760120.1 | XP_008758342.1 | 1 - 2 | 168 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl21
Genomic Map