Assay Details
| Gene Symbol: Kat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005872.1 | NP_001005872.1 | 7 - 8 | 938 | 106 | |
| GenBank mRNA | AF333984.1 | - | 5 - 6 | 591 | 106 | |
| BC083879.1 | - | 7 - 8 | 938 | 7130366 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kat5
Genomic Map