Assay Details
| Gene Symbol: Ano1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107564.1 | NP_001101034.1 | 2 - 3 | 330 | 61 | |
| XM_006230780.2 | XP_006230842.1 | 2 - 3 | 398 | 61 | ||
| XM_008760118.1 | XP_008758340.1 | 2 - 3 | 399 | 61 | ||
| XM_008760119.1 | XP_008758341.1 | 2 - 3 | 316 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ano1
Genomic Map