Assay Details
| Gene Symbol: Mtl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012069.2 | NP_001012069.2 | 5 - 6 | 869 | 92 | |
| GenBank mRNA | BC083920.1 | - | 5 - 6 | 855 | 7135090 | 92 |
More Information
Updated Mapping Information
Set Membership: |
|

Mtl5
Genomic Map