Assay Details
| Gene Symbol: Mina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768680.1 | XP_008766902.1 | 1 - 2 | 87 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768680.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mina
Genomic Map