Assay Details
| Gene Symbol: Cep19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768696.1 | XP_008766918.1 | 1 - 2 | 321 | 109 | |
| XM_008768697.1 | XP_008766919.1 | 1 - 2 | 322 | 109 | ||
| GenBank mRNA | FQ211926.1 | - | 1 - 2 | 159 | 109 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_008768696.1,XM_008768697.1 transcripts.
This assay is in the 5'UTR region of the XM_008768696.1,XM_008768697.1 transcripts.
Set Membership: |
|

Cep19
Genomic Map