Assay Details
Gene Symbol: Ppp1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138823.2 | NP_620178.1 | 250 | 96 | ||
GenBank mRNA | BC092194.1 | - | 250 | 7313160 | 96 | |
S79213.1 | - | 265 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
