Assay Details
| Gene Symbol: Iqcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107092.1 | NP_001100562.1 | 4 - 5 | 383 | 68 | |
| XM_006248395.2 | XP_006248457.1 | 4 - 5 | 449 | 68 | ||
| XM_006248396.2 | XP_006248458.1 | 4 - 5 | 454 | 68 | ||
| XM_006248397.2 | XP_006248459.1 | 4 - 5 | 417 | 68 | ||
| XM_008768730.1 | XP_008766952.1 | 4 - 5 | 422 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Iqcb1
Genomic Map