Assay Details
| Gene Symbol: Cd80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012926.1 | NP_037058.1 | 3 - 4 | 729 | 81 | |
| XM_006248365.2 | XP_006248427.1 | 3 - 4 | 81 | |||
| GenBank mRNA | AF010465.1 | - | 3 - 4 | 729 | 81 | |
| U10925.1 | - | 3 - 4 | 81 | |||
| U88622.1 | - | 3 - 4 | 81 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cd80
Genomic Map