Assay Details
| Gene Symbol: Sidt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100653.1 | NP_001094123.1 | 7 - 8 | 836 | 69 | |
| XM_006248328.2 | XP_006248390.1 | 7 - 8 | 1718 | 69 | ||
| XM_008768718.1 | XP_008766940.1 | 7 - 8 | 1717 | 69 | ||
| GenBank mRNA | AY562215.1 | - | 7 - 8 | 836 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sidt1
Genomic Map