Assay Details
| Gene Symbol: Wdr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024766.1 | NP_001019937.1 | 1 | 1702 | 163 | |
| GenBank mRNA | BC097449.1 | - | 1 | 1702 | 7456711 | 163 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Wdr5b
Genomic Map