Assay Details
Gene Symbol: Fbxo40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248404.2 | XP_006248466.1 | 2 - 3 | 309 | 143 | |
XM_344023.7 | XP_344024.5 | 1 - 2 | 194 | 143 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006248404.2, XM_344023.7 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
