Assay Details
| Gene Symbol: Pdia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014125.1 | NP_001014147.1 | 14 - 15 | 1300 | 67 | |
| GenBank mRNA | BC088305.1 | - | 14 - 15 | 1300 | 7323988 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P993914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117489 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P993914 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P993914 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P993914 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P993914 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8600 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50251 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83045 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89138 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5412 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63565 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23151 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10751507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751512 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751501 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751503 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751502 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751505 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751500 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751504 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751509 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751499 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10751498 | - | - |
| Rat Genome 230 2.0 Array | 1374828_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374828_at | - | - |
| Affymetrix HT Rat Focus Array | 1374828_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374828_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100003854.rn.1 | TC1100000705.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374828_PM_at | - | - |
| Rat Expression Set 230 | 1374828_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751498 | - | - |
| Rat Genome U34 Set | rc_AI045590_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20827380 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350619 | - | - |

Pdia5
Genomic Map