Assay Details
| Gene Symbol: LOC100911928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248418.2 | XP_006248480.1 | 3 - 4 | 374 | 156 | |
| Gene Symbol: Pdia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014125.1 | NP_001014147.1 | 3 - 4 | 283 | 156 | |
| GenBank mRNA | BC088305.1 | - | 3 - 4 | 283 | 7323988 | 156 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911928
Genomic Map