Assay Details
Gene Symbol: RGD1564541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107950.1 | NP_001101420.1 | 1 - 2 | 78 | 99 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001107950.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
