Assay Details
Gene Symbol: RGD1564541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107950.1 | NP_001101420.1 | 7 - 8 | 865 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P272186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178733 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P272186 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P272186 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P272186 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P272186 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102710 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71759 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10932035 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932034 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932031 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932030 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932032 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932033 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932036 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10932029 | - | - |
Rat Genome 230 2.0 Array | 1380185_at | - | - |
Rat Genome 230 2.0 Array | 1394891_at | - | - |
Rat Genome 230 2.0 Array | 1394891_PM_at | - | - |
Rat Genome 230 2.0 Array | 1380185_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00014695.rn.1 | TC0X00002351.rn.1 | RTA TC |
Rat Expression Set 230 | 1380185_at | - | - |
Rat Expression Set 230 | 1380185_PM_at | - | - |
Rat Expression Set 230 | 1394891_at | - | - |
Rat Expression Set 230 | 1394891_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10932029 | - | - |
Rat Genome U34 Set | rc_AI045223_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20717161 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363965 | - | - |
