Assay Details
| Gene Symbol: Rpl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031102.2 | NP_112364.1 | 4 - 5 | 316 | 86 | |
| XM_006229161.2 | XP_006229223.1 | 4 - 5 | 797 | 86 | ||
| GenBank mRNA | BC084727.1 | - | 3 - 4 | 308 | 7308070 | 86 |
| CB577548.1 | - | 4 - 5 | 316 | 86 | ||
| DQ403026.1 | - | 3 - 4 | 232 | 86 | ||
| FQ211765.1 | - | 4 - 5 | 334 | 86 | ||
| FQ221110.1 | - | 4 - 5 | 334 | 86 | ||
| FQ222651.1 | - | 4 - 5 | 334 | 86 | ||
| FQ228615.1 | - | 4 - 5 | 337 | 86 | ||
| M20156.1 | - | 3 - 4 | 302 | 86 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl18
Genomic Map