Assay Details
| Gene Symbol: Slc6a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203334.1 | NP_976079.1 | 15 - 16 | 2501 | 87 | |
| XM_008759310.1 | XP_008757532.1 | 15 - 16 | 3312 | 87 | ||
| XM_008759311.1 | XP_008757533.1 | 16 - 17 | 2510 | 87 | ||
| GenBank mRNA | AY547309.1 | - | 15 - 16 | 2393 | 87 | |
| L21672.2 | - | 15 - 16 | 2501 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P060942 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P060947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154684 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P060942 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P060947 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P060947 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P060942 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P060947 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P060942 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P226649 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67463 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13539 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90337 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10707264 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707260 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707263 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707257 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707259 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707265 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707258 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707267 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707261 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707266 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707262 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10707256 | - | - |
| Rat Genome 230 2.0 Array | 1371047_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371047_at | - | - |
| Affymetrix HT Rat Focus Array | 1371047_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371047_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100012627.rn.1 | TC0100001560.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371047_at | - | - |
| Rat Expression Set 230 | 1371047_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10707256 | - | - |
| Rat Genome U34 Set | L21672_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21652569 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374850 | - | - |
| RatRef-12 Expression BeadChip | ILMN_52767 | - | - |

Slc6a5
Genomic Map