Assay Details
| Gene Symbol: Plekhf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013148.1 | NP_001013166.1 | 1 | 541 | 91 | |
| XM_006228905.2 | XP_006228967.1 | 2 | 636 | 91 | ||
| GenBank mRNA | BC079354.1 | - | 1 | 541 | 7108247 | 91 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Plekhf1
Genomic Map