Assay Details
| Gene Symbol: Nomo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108484.1 | NP_001101954.1 | 1 - 2 | 209 | 61 | |
| XM_006229108.2 | XP_006229170.1 | 1 - 2 | 224 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nomo1
Genomic Map