Assay Details
Gene Symbol: Rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008694.2 | NP_001008694.1 | 1 - 2 | 103 | 81 | |
GenBank mRNA | BC083719.1 | - | 1 - 2 | 104 | 7190838 | 81 |
CK601329.1 | - | 1 - 2 | 103 | 81 | ||
FQ228883.1 | - | 1 - 2 | 124 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC083719.1,NM_001008694.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
