Assay Details
| Gene Symbol: Ap2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107511.2 | NP_001100981.1 | 6 - 7 | 710 | 74 | |
| XM_006229062.2 | XP_006229124.1 | 5 - 6 | 2554 | 74 | ||
| XM_006229063.2 | XP_006229125.1 | 5 - 6 | 2554 | 74 | ||
| XM_006229064.2 | XP_006229126.1 | 6 - 7 | 818 | 74 | ||
| XM_006229065.2 | XP_006229127.1 | 6 - 7 | 818 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ap2a1
Genomic Map